missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylenediamine tetraacetic acid disodium salt dihydrate, 99.74%, MP Biomedicals™
$104.64 - $327.54
Chemical Identifiers
| CAS | 6381-92-6 |
|---|---|
| Molecular Formula | C10H18N2Na2O10 |
| Molecular Weight (g/mol) | 372.24 |
| MDL Number | MFCD00150037,MFCD00003541 |
| InChI Key | OVBJJZOQPCKUOR-UHFFFAOYSA-L |
| Synonym | edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs |
| PubChem CID | 44120005 |
| IUPAC Name | disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate |
| SMILES | O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN19517380
|
MP Biomedicals Inc
0219517380 |
100 g |
Each for $104.64
|
|
|||||
|
ICN19517383
|
MP Biomedicals Inc
0219517383 |
250 g |
Each for $184.36
|
|
|||||
|
ICN19517390
|
MP Biomedicals Inc
0219517390 |
500 g |
Each for $245.22
|
|
|||||
|
ICN19517391
|
MP Biomedicals Inc
0219517391 |
1 kg |
Each for $327.54
|
|
|||||
Chemical Identifiers
| 6381-92-6 | |
| 372.24 | |
| OVBJJZOQPCKUOR-UHFFFAOYSA-L | |
| 44120005 | |
| O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O |
| C10H18N2Na2O10 | |
| MFCD00150037,MFCD00003541 | |
| edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs | |
| disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate |
Specifications
| 6381-92-6 | |
| C10H18N2Na2O10 | |
| 100 g | |
| OVBJJZOQPCKUOR-UHFFFAOYSA-L | |
| disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate | |
| 44120005 | |
| 0.9974 | |
| Ethylenediamine tetraacetic acid disodium salt dihydrate |
| White | |
| MFCD00150037,MFCD00003541 | |
| edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs | |
| O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O | |
| 372.24 | |
| 372.2 | |
| Powder |