missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethylenediamine tetraacetic acid disodium salt dihydrate, 99.74%, MP Biomedicals™
Manufacturer: Mp Biomedicals 0219517380
Specifications
Ethylenediamine tetraacetic acid disodium salt dihydrate | |
White | |
MFCD00150037,MFCD00003541 | |
100g | |
O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O | |
372.24 | |
372.2 | |
0.9974 |
6381-92-6 | |
C10H18N2Na2O10 | |
buffer solution, ph 10.00, cal-ex decalcifier, edta disodium salt, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs, sodium di ethylenediamine tetraacetate dihydrate, sodium di ethylenediamine tetraacetate standard solution | |
OVBJJZOQPCKUOR-UHFFFAOYSA-L | |
disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate | |
44120005 | |
Powder |
Chemical Identifiers
6381-92-6 | |
372.24 | |
OVBJJZOQPCKUOR-UHFFFAOYSA-L | |
44120005 | |
O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O |
C10H18N2Na2O10 | |
MFCD00150037,MFCD00003541 | |
buffer solution, ph 10.00, cal-ex decalcifier, edta disodium salt, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs, sodium di ethylenediamine tetraacetate dihydrate, sodium di ethylenediamine tetraacetate standard solution | |
disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate |