missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediamine Sulfate 98.0+%, TCI America™

Catalog No. E008225G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E008225G 25 g
E0082500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E008225G Supplier TCI America Supplier No. E008225G

Chemical Identifiers

CAS 22029-36-3
Molecular Formula C2H10N2O4S
Molecular Weight (g/mol) 158.17
MDL Number MFCD00036417
InChI Key BNZCDZDLTIHJAC-UHFFFAOYSA-N
PubChem CID 168015
IUPAC Name ethane-1,2-diamine; sulfuric acid
SMILES NCCN.OS(O)(=O)=O

Specifications

Color White
Quantity 25 g
UN Number 2811
Formula Weight 158.17
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Ethylenediamine Sulfate
EINECSNumber (2)-0150
RTECSNumber KV5425000
TSCA Yes