missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ethylenediamine-N,N'-diacetic acid

Catalog No. AAJ6357014
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6357014 25 g
AAJ6357006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6357014 Supplier Thermo Scientific Chemicals Supplier No. J6357014
Only null left

Acts as a chelating agent | CAS: 5657-17-0 | C6H10N2Na2O4 | 220.14 g/mol

Ethylenediamine-N,N'-diacetic acid is acts as a chelating agent, used as a haemostatic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethylenediamine-N,N′-diacetic acid is acts as a chelating agent, used as a haemostatic.

Solubility
Soluble in warm water.

Notes
Protect from heat. Store away from oxidizing agents.

Chemical Identifiers

CAS 5657-17-0
Molecular Formula C6H10N2Na2O4
Molecular Weight (g/mol) 220.14
MDL Number MFCD00004281
InChI Key QMIFISRIAJBCGZ-UHFFFAOYSA-L
Synonym edda, ethylenediamine-n,n'-diacetic acid, n,n'-ethylenediglycine, glycine, n,n'-1,2-ethanediylbis, ethylenediaminediacetic acid, glycine, n,n'-ethylenedi, unii-4lx0u5342u, 2-2-carboxymethylamino ethylamino acetic acid, ethylenediamine-n,n'-diaceticacid, ethylenediamine-n,n'-di acetic acid
PubChem CID 61975
SMILES [Na+].[Na+].[O-]C(=O)CNCCNCC([O-])=O

Specifications

Melting Point 224°C
Color White
Linear Formula HOOCCH2NHCH2CH2NHCH2COOH
Quantity 25 g
Beilstein 1778355
Solubility Information Soluble in warm water.
IUPAC Name disodium 2-({2-[(carboxylatomethyl)amino]ethyl}amino)acetate
Formula Weight 176.17
Physical Form Powder
Chemical Name or Material Ethylenediamine-N,N'-diacetic acid
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 227-105-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only