missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediamine Dihydrochloride 98.0+%, TCI America™

Catalog No. p-7128827
Change view
Click to view available options
Quantity:
25 g
500 g
Catalog No. Quantity
E007825G 25 g
E0078500G 500 g
2 options

Catalog No. E007825G

Supplier: TCI America E007825G

Only null left

Chemical Identifiers

CAS 333-18-6
Molecular Formula C2H10Cl2N2
Molecular Weight (g/mol) 133.02
MDL Number MFCD00012524
InChI Key OHHBFEVZJLBKEH-UHFFFAOYSA-N
Synonym ethylenediamine dihydrochloride, chlor-ethamine, ethylenediammonium chloride, ethylenediammonium dichloride, ethane-1,2-diamine dihydrochloride, dimethylenediamine dihydrochloride, 1,2-diaminoethane dihydrochloride, 1,2-ethanediamine, dihydrochloride, ethylenediamine 2hcl, dimethylenediamine diydrochloride
PubChem CID 9521
ChEBI CHEBI:53626
IUPAC Name dihydrogen ethane-1,2-diamine dichloride
SMILES [H+].[H+].[Cl-].[Cl-].NCCN

Specifications

Color White
Quantity 25 g
Formula Weight 133.02
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Ethylenediamine Dihydrochloride
EINECSNumber (2)-0150
RTECSNumber KV3850000
TSCA Yes