accessibility menu, dialog, popup

UserName

HHClClNH2H2N

CAS RN 333-18-6

IUPAC Name: dihydrogen ethane-1,2-diamine dichloride
Synonyms: dihydrogen ethylenediamine dichloride
Molecular Weight (g/mol): 133.02
Molecular Formula: C2H10Cl2N2
InChi Key: OHHBFEVZJLBKEH-UHFFFAOYSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].NCCN
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (5)