Learn More
Ethylenediamine dihydrochloride, 98%
CAS: 333-18-6 | C2H10Cl2N2 | 133.02 g/mol
Supplier: Thermo Scientific Chemicals A135630E
Description
Ethylenediamine dihydrochloride was used in modified ethylenediamine condensation method for fluorimetric determination of catecholamines. It was used to investigate the luminescence properties of complexes of EuIII and TbIII with ethylenediamine. It was used in the synthesis of 1,3,5-tris(4,5-dihydro-1H-imidazol-2-yl)benzene3
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthylenediamine dihydrochloride was used in modified ethylenediamine condensation method for fluorimetric determination of catecholamines. It was used to investigate the luminescence properties of complexes of EuIII and TbIII with ethylenediamine. It was used in the synthesis of 1,3,5-tris(4,5-dihydro-1H-imidazol-2-yl)benzene3
Solubility
Soluble in water 300 g/L (20°C).
Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
| Ethylenediamine dihydrochloride | |
| >300°C | |
| MFCD00012524 | |
| Hygroscopic | |
| Soluble in water 300g/L (20°C). | |
| [H+].[H+].[Cl-].[Cl-].NCCN | |
| 133.02 | |
| CHEBI:53626 | |
| 98% |
| 333-18-6 | |
| C2H10Cl2N2 | |
| 2500 g | |
| ethylenediamine dihydrochloride, chlor-ethamine, ethylenediammonium chloride, ethylenediammonium dichloride, ethane-1,2-diamine dihydrochloride, dimethylenediamine dihydrochloride, 1,2-diaminoethane dihydrochloride, 1,2-ethanediamine, dihydrochloride, ethylenediamine 2hcl, dimethylenediamine diydrochloride | |
| OHHBFEVZJLBKEH-UHFFFAOYSA-N | |
| ethane-1,2-diamine;dihydrochloride | |
| 9521 | |
| 133.02 |
Chemical Identifiers
| 333-18-6 | |
| 133.02 | |
| OHHBFEVZJLBKEH-UHFFFAOYSA-N | |
| 9521 | |
| ethane-1,2-diamine;dihydrochloride |
| C2H10Cl2N2 | |
| MFCD00012524 | |
| ethylenediamine dihydrochloride, chlor-ethamine, ethylenediammonium chloride, ethylenediammonium dichloride, ethane-1,2-diamine dihydrochloride, dimethylenediamine dihydrochloride, 1,2-diaminoethane dihydrochloride, 1,2-ethanediamine, dihydrochloride, ethylenediamine 2hcl, dimethylenediamine diydrochloride | |
| CHEBI:53626 | |
| [H+].[H+].[Cl-].[Cl-].NCCN |
Safety and Handling
GHS H Statement
H361-H373-H302
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
P261-P264b-P270-P271-P272-P280-P285-P301+P312-P302+P352-P304+P341-P305+P351+P338-P312-P330-P333+P313-P342+P311-P362-P501c
H302-H315-H317-H319-H334-H335
EINECSNumber : 206-369-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only