missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediamine dihydrochloride, 98%

Catalog No. AAA1356336
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1356336 500 g
AAA1356322 100 g
AAA135630E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1356336 Supplier Thermo Scientific Chemicals Supplier No. A1356336
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 333-18-6 | C2H10Cl2N2 | 133.02 g/mol

Ethylenediamine dihydrochloride was used in modified ethylenediamine condensation method for fluorimetric determination of catecholamines. It was used to investigate the luminescence properties of complexes of EuIII and TbIII with ethylenediamine. It was used in the synthesis of 1,3,5-tris(4,5-dihydro-1H-imidazol-2-yl)benzene3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethylenediamine dihydrochloride was used in modified ethylenediamine condensation method for fluorimetric determination of catecholamines. It was used to investigate the luminescence properties of complexes of EuIII and TbIII with ethylenediamine. It was used in the synthesis of 1,3,5-tris(4,5-dihydro-1H-imidazol-2-yl)benzene3

Solubility
Soluble in water 300 g/L (20°C).

Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 333-18-6
Molecular Formula C2H10Cl2N2
Molecular Weight (g/mol) 133.02
MDL Number MFCD00012524
InChI Key OHHBFEVZJLBKEH-UHFFFAOYSA-N
Synonym ethylenediamine dihydrochloride, chlor-ethamine, ethylenediammonium chloride, ethylenediammonium dichloride, ethane-1,2-diamine dihydrochloride, dimethylenediamine dihydrochloride, 1,2-diaminoethane dihydrochloride, 1,2-ethanediamine, dihydrochloride, ethylenediamine 2hcl, dimethylenediamine diydrochloride
PubChem CID 9521
ChEBI CHEBI:53626
IUPAC Name ethane-1,2-diamine;dihydrochloride
SMILES [H+].[H+].[Cl-].[Cl-].NCCN

Specifications

Melting Point >300°C
Quantity 500 g
Sensitivity Hygroscopic
Solubility Information Soluble in water 300g/L (20°C).
Formula Weight 133.02
Percent Purity 98%
Chemical Name or Material Ethylenediamine dihydrochloride
Hazard Category H302-H315-H317-H319-H334-H335
Hazard Statement GHS H Statement
H361-H373-H302
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P285-P301+P312-P302+P352-P304+P341-P305+P351+P338-P312-P330-P333+P313-P342+P311-P362-P501c
EINECSNumber 206-369-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only