missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediamine Dihydrobromide 97.0+%, TCI America™

Catalog No. E12215G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E12215G 5 g
E12211G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E12215G Supplier TCI America Supplier No. E12215G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 624-59-9
Molecular Formula C2H10Br2N2
Molecular Weight (g/mol) 221.92
MDL Number MFCD07783256
InChI Key BCQZYUOYVLJOPE-UHFFFAOYSA-N
PubChem CID 164699
IUPAC Name dihydrogen ethane-1,2-diamine dibromide
SMILES [H+].[H+].[Br-].[Br-].NCCN

Specifications

Melting Point 350°C
Quantity 5 g
Formula Weight 221.92
Percent Purity ≥97.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material Ethylenediamine Dihydrobromide