accessibility menu, dialog, popup

UserName

HHBrBrNH2H2N

CAS RN 624-59-9

IUPAC Name: dihydrogen ethane-1,2-diamine dibromide
Synonyms: dihydrogen ethylenediamine dibromide
Molecular Weight (g/mol): 221.92
Molecular Formula: C2H10Br2N2
InChi Key: BCQZYUOYVLJOPE-UHFFFAOYSA-N
SMILES: [H+].[H+].[Br-].[Br-].NCCN
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results