missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediamine, 70%, Technical Grade, Ricca Chemical

Catalog No. rsoe01001a
Change view
Click to view available options
Quantity:
1 L
Packaging:
Natural Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RSOE01001A 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RSOE01001A Supplier Ricca Chemical Company Supplier No. RSOE01001A
Only null left

Chemical Identifiers

CAS 107-15-3 , 7732-18-5
Molecular Formula C2H8N2
Molecular Weight (g/mol) Mixture
MDL Number MFCD00008204
InChI Key PIICEJLVQHRZGT-UHFFFAOYSA-N
Synonym ethylenediamine, 1,2-ethanediamine, 1,2-diaminoethane, ethylene diamine, ethylenediamine, dimethylenediamine, edamine, 1,2-ethylenediamine, aethaldiamin, aethylenediamin
PubChem CID 3301
ChEBI CHEBI:30347
IUPAC Name ethane-1,2-diamine
SMILES NCCN

Specifications

CAS Min % 29.0, 69.0
CAS Max % 31.0, 71.0
Color Yellow
Boiling Point Approximately 117°C
Packaging Natural Poly Bottle
Quantity 1 L
Percent Purity 70% w/w
Grade Technical
Concentration or Composition (by Analyte or Components) 70%
Physical Form Liquid
Chemical Name or Material Ethylenediamine
Show More Show Less
ShelfLife 60 months