missing translation for 'onlineSavingsMsg'
Learn More

Ethylenebis(triphenylphosphine)platinum(0), 98%

CAS: 12120-15-9 | C38H34P2Pt | 747.72 g/mol

$272.27 - $272.27

Chemical Identifiers

CAS 12120-15-9
Molecular Formula C38H34P2Pt
Molecular Weight (g/mol) 747.72
MDL Number MFCD00012028
InChI Key RZUASTIKPBCXPU-UHFFFAOYSA-N
IUPAC Name ethene bis(triphenylphosphane) platinum
SMILES [Pt].C=C.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC369362500
SDS View Documents
Thermo Scientific Chemicals
369362500
250 mg Glass bottle
Each for $272.27
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12120-15-9
747.72
RZUASTIKPBCXPU-UHFFFAOYSA-N
[Pt].C=C.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
C38H34P2Pt
MFCD00012028
ethene bis(triphenylphosphane) platinum

Specifications

12120-15-9
97.5% min. (ex Pt)
C38H34P2Pt
MFCD00012028
Solubility in water: insoluble
[Pt].C=C.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
747.72
98%
127.0°C to 131.0°C
Glass bottle
[(C6H5)3P]2Pt[H2C=CH2]
250 mg
RZUASTIKPBCXPU-UHFFFAOYSA-N
ethene bis(triphenylphosphane) platinum
747.73
Ethylenebis(triphenylphosphine)platinum(0)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only