accessibility menu, dialog, popup

UserName

PPtPPhPhPhPhPhPh

CAS RN 12120-15-9

IUPAC Name: ethene bis(triphenylphosphane) platinum
Synonyms: ethylene bis(triphenylphosphine) platinum
Molecular Weight (g/mol): 747.72
Molecular Formula: C38H34P2Pt
InChi Key: RZUASTIKPBCXPU-UHFFFAOYSA-N
SMILES: [Pt].C=C.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results