missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Trithiocarbonate 96.0+%, TCI America™

$303.84 - $987.95

Chemical Identifiers

CAS 822-38-8
Molecular Formula C3H4S3
Molecular Weight (g/mol) 136.245
MDL Number MFCD00005493
InChI Key XCWPBWWTGHQKDR-UHFFFAOYSA-N
Synonym ethylene trithiocarbonate, cyclic ethylene trithiocarbonate, trithiocarbonic acid, cyclic ethylene ester, carbonic acid, trithio-, cyclic ethylene ester, cylic ethylene trithiocarbonate, acmc-209pnw, 1,3-dithiolan-2-thione, 5-19-04-00014 beilstein handbook reference, ksc449a8r
PubChem CID 13196
IUPAC Name 1,3-dithiolane-2-thione
SMILES C1CSC(=S)S1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
E032325G
View Documents
TCI America
E032325G
25 g N/A
Only null left
 
E0323100G
View Documents
TCI America
E0323100G
100 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

822-38-8
136.245
XCWPBWWTGHQKDR-UHFFFAOYSA-N
13196
C1CSC(=S)S1
C3H4S3
MFCD00005493
ethylene trithiocarbonate, cyclic ethylene trithiocarbonate, trithiocarbonic acid, cyclic ethylene ester, carbonic acid, trithio-, cyclic ethylene ester, cylic ethylene trithiocarbonate, acmc-209pnw, 1,3-dithiolan-2-thione, 5-19-04-00014 beilstein handbook reference, ksc449a8r
1,3-dithiolane-2-thione

Specifications

822-38-8
MFCD00005493
ethylene trithiocarbonate, cyclic ethylene trithiocarbonate, trithiocarbonic acid, cyclic ethylene ester, carbonic acid, trithio-, cyclic ethylene ester, cylic ethylene trithiocarbonate, acmc-209pnw, 1,3-dithiolan-2-thione, 5-19-04-00014 beilstein handbook reference, ksc449a8r
C1CSC(=S)S1
136.245
136.25
Crystals
C3H4S3
25 g
XCWPBWWTGHQKDR-UHFFFAOYSA-N
1,3-dithiolane-2-thione
13196
≥96.0% (GC)
Ethylene Trithiocarbonate
Videos
Safety and Handling

Safety and Handling

RTECSNumber : FG2720000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'