accessibility menu, dialog, popup

UserName

SSS

CAS RN 822-38-8

IUPAC Name: 1,3-dithiolane-2-thione
Synonyms: 1,3-dithiolane-2-thione
Molecular Weight (g/mol): 136.25
Molecular Formula: C3H4S3
InChi Key: XCWPBWWTGHQKDR-UHFFFAOYSA-N
SMILES: S=C1SCCS1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results