missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Trithiocarbonate 96.0+%, TCI America™

Catalog No. E032325G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E032325G 25 g
E0323100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E032325G Supplier TCI America Supplier No. E032325G
Only null left

Chemical Identifiers

CAS 822-38-8
Molecular Formula C3H4S3
Molecular Weight (g/mol) 136.245
MDL Number MFCD00005493
InChI Key XCWPBWWTGHQKDR-UHFFFAOYSA-N
Synonym ethylene trithiocarbonate, cyclic ethylene trithiocarbonate, trithiocarbonic acid, cyclic ethylene ester, carbonic acid, trithio-, cyclic ethylene ester, cylic ethylene trithiocarbonate, acmc-209pnw, 1,3-dithiolan-2-thione, 5-19-04-00014 beilstein handbook reference, ksc449a8r
PubChem CID 13196
IUPAC Name 1,3-dithiolane-2-thione
SMILES C1CSC(=S)S1

Specifications

Quantity 25 g
Formula Weight 136.25
Percent Purity ≥96.0% (GC)
Physical Form Crystals
Chemical Name or Material Ethylene Trithiocarbonate
RTECSNumber FG2720000
TSCA Yes