missing translation for 'onlineSavingsMsg'
Learn More

Ethylene glycol, technical

Catalog No. AC444230010
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC444230010 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC444230010 Supplier Thermo Scientific Chemicals Supplier No. 444230010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 107-21-1 | C2H6O2 | 62.068 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 107-21-1
Molecular Formula C2H6O2
Molecular Weight (g/mol) 62.068
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
PubChem CID 174
ChEBI CHEBI:30742
IUPAC Name ethane-1,2-diol
SMILES C(CO)O

Specifications

Melting Point -13.0°C
Density 1.1130g/mL
Quantity 1 L
Flash Point 116°C
Refractive Index 1.4300 to 1.4330
Specific Gravity 1.113
Linear Formula HOCH2CH2OH
Viscosity 21 mPa.s (20°C)
Water 0.2% max.
Boiling Point 196.0°C to 198.0°C
Solubility Information Solubility in water: miscible. Other solubilities: miscible with acetic acid, acetone, glycerol,, pyridine, lower aliphatic alcohols,, aldehydes and ketones, slightly soluble in ether, practically insoluble in benzene and hydrocarbons
Formula Weight 62.06
Percent Purity ≥98.5% (GC)
Grade Technical
Chemical Name or Material Ethylene glycol
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H StatementHarmful if swallowed.May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement GHS P StatementWear protective gloves/protective clothing/eye protection/face protection.IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

RUO – Research Use Only