missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylene Glycol 99.5+%, TCI America™
$43.67 - $55.07
Chemical Identifiers
| CAS | 107-21-1 |
|---|---|
| Molecular Formula | C2H6O2 |
| Molecular Weight (g/mol) | 62.068 |
| MDL Number | MFCD00002885 |
| InChI Key | LYCAIKOWRPUZTN-UHFFFAOYSA-N |
| Synonym | ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol |
| PubChem CID | 174 |
| ChEBI | CHEBI:30742 |
| IUPAC Name | ethane-1,2-diol |
| SMILES | C(CO)O |
Chemical Identifiers
| 107-21-1 | |
| 62.068 | |
| LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
| 174 | |
| ethane-1,2-diol |
| C2H6O2 | |
| MFCD00002885 | |
| ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol | |
| CHEBI:30742 | |
| C(CO)O |
Specifications
| Clear Liquid at 20°C | |
| 196°C | |
| Colorless | |
| C2H6O2 | |
| ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol | |
| C(CO)O | |
| 62.068 | |
| CHEBI:30742 | |
| ≥99.5% (GC) |
| -13°C | |
| 107-21-1 | |
| 25 g | |
| MFCD00002885 | |
| LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
| ethane-1,2-diol | |
| 174 | |
| 62.07 | |
| Ethylene Glycol |
Safety and Handling
EINECSNumber : (2)-0230
RTECSNumber : KW2975000
TSCA : Yes