missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Glycol 99.5+%, TCI America™

Catalog No. E0105500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0105500G 500 g
E010525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E0105500G Supplier TCI America Supplier No. E0105500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 107-21-1
Molecular Formula C2H6O2
Molecular Weight (g/mol) 62.068
MDL Number MFCD00002885
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
PubChem CID 174
ChEBI CHEBI:30742
IUPAC Name ethane-1,2-diol
SMILES C(CO)O

Specifications

Physical Form Clear Liquid at 20°C
Melting Point -13°C
Boiling Point 196°C
Color Colorless
Quantity 500 g
Formula Weight 62.07
Percent Purity ≥99.5% (GC)
Chemical Name or Material Ethylene Glycol
EINECSNumber (2)-0230
RTECSNumber KW2975000
TSCA Yes