missing translation for 'onlineSavingsMsg'
Learn More

Ethylene glycol Solution, NMR Reference Standard, 80% in DMSO-d6 (99.9 atom% D), NMR tube size 5 mm 8 in., MilliporeSigma™ Supelco™

Catalog No. 111012407
Change view
Click to view available options
Quantity:
0.7 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012407 0.7 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012407 Supplier Merck Emd Millipore Supplier No. 7219728IN

Specifications

Packaging 5 mm O.D. tube
CAS 107-21-1
Linear Formula C2H6O2
MDL Number MFCD00002885
Type NMR Reference Standard
Quantity 0.7 mL
Formula Weight 62.07
Grade NMR Reference Standard
Concentration or Composition (by Analyte or Components) 80% in DMSO- d6 (99.9 atom % D)