missing translation for 'onlineSavingsMsg'
Learn More

Ethylene glycol, Reagent Grade, ≥99%, Honeywell™

Catalog No. p-7121613
Change view
Click to view available options
Quantity:
1 L
5 L
20 L
30 kg
4 x 2.5 L
Packaging:
Poly Bottle
Stainless Steel Can
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60044640 1 L Poly Bottle
60044647 5 L Poly Bottle
60044645 4 x 2.5 L Poly Bottle
60044642 20 L Stainless Steel Can
60044644 30 kg Stainless Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 60044640 Supplier Honeywell Chemicals Supplier No. 1024661L
Only null left

Reagent Grade,≥99%

  • ≥95% for general laboratory use.

Chemical Identifiers

CAS 107-21-1
Molecular Formula C2H6O2
Molecular Weight (g/mol) 62.068
MDL Number MFCD00002885
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
PubChem CID 174
ChEBI CHEBI:30742
IUPAC Name ethane-1,2-diol
SMILES C(CO)O

Specifications

Melting Point -13°C (literature)
CAS Max % 100.0000%
Density 1.113 g/mL at 25 °C(lit.)
Quantity 1 L
Flash Point 111°C
Packaging Poly Bottle
Linear Formula HOCH2CH2OH
Beilstein 505945
Boiling Point 197°C
Formula Weight 62.07
Percent Purity >99%
Grade Reagent
Chemical Name or Material Ethylene glycol, Reagent Grade, ≥99%
Show More Show Less
ShelfLife 1080 days from date of manufacture
EINECSNumber 203-473-3
Recommended Storage Room Temp