missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Glycol, Reagent, 99%, Spectrum™ Chemical

Catalog No. 18602568
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
6 x 500 mL
20 L
4 x 4 L
6 x 1 L
55 gal.
Packaging:
Amber Glass Bottle
Drum
Poly Bottle
Poly Drum
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602568 55 gal. Drum
18602559 500 mL Poly Bottle
18602561 1 L Poly Bottle
18602560 6 x 500 mL Poly Bottle
18602562 4 L Amber Glass Bottle
18602563 4 L Poly Bottle
18602564 6 x 1 L Poly Bottle
18602565 4 x 4 L Amber Glass Bottle
18602566 4 x 4 L Poly Bottle
18602567 20 L Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 18602568 Supplier Spectrum Chemical Mfg Cor Supplier No. E105155GLBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

E1051, 107-21-1, C2H6O2

Spectrum™ Chemical Ethylene Glycol, Reagent is a colorless, odorless, sweet tasting organic compound that has a syrupy texture and is a liquid. It has many lab applications including use as an antifreeze, use as a desiccant, protecting group for carbonyls during chemical synthesis and as a preservative for biological specimens. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.

Chemical Identifiers

CAS 107-21-1
Molecular Formula C2H6O2
Molecular Weight (g/mol) 62.07
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
IUPAC Name ethane-1,2-diol
SMILES OCCO

Specifications

Acidity 0.005%
CAS Max % 100%
Quantity 55 gal.
Packaging Drum
Boiling Point 196°C to 199°C
Percent Purity 99%
Grade Reagent
Chemical Name or Material Ethylene Glycol