missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Glycol, Reagent, 99%, Spectrum™ Chemical

Catalog No. 18602566
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 L
20 L
4 L
4 x 4 L
500 mL
55 gal.
6 x 1 L
6 x 500 mL
Packaging:
Amber Glass Bottle
Drum
Poly Bottle
Poly Drum
Catalog No. Quantity Packaging
18602566 4 x 4 L Poly Bottle
18602559 500 mL Poly Bottle
18602561 1 L Poly Bottle
18602560 6 x 500 mL Poly Bottle
18602562 4 L Amber Glass Bottle
18602563 4 L Poly Bottle
18602564 6 x 1 L Poly Bottle
18602565 4 x 4 L Amber Glass Bottle
18602567 20 L Poly Drum
18602568 55 gal. Drum
10 options

Catalog No. 18602566

Supplier: Spectrum Chemical Mfg Cor E10514LTPLCS4

Only null left

E1051, 107-21-1, C2H6O2

Spectrum™ Chemical Ethylene Glycol, Reagent is a colorless, odorless, sweet tasting organic compound that has a syrupy texture and is a liquid. It has many lab applications including use as an antifreeze, use as a desiccant, protecting group for carbonyls during chemical synthesis and as a preservative for biological specimens. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.

Chemical Identifiers

CAS 107-21-1
Molecular Formula C2H6O2
Molecular Weight (g/mol) 62.07
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
IUPAC Name ethane-1,2-diol
SMILES OCCO

Specifications

Acidity 0.005%
CAS Max % 100%
Boiling Point 196°C to 199°C
Quantity 4 x 4 L
Packaging Poly Bottle
Percent Purity 99%
Grade Reagent
Chemical Name or Material Ethylene Glycol