missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylene Glycol, Reagent, 99%, Spectrum™ Chemical
Description
Spectrum™ Chemical Ethylene Glycol, Reagent is a colorless, odorless, sweet tasting organic compound that has a syrupy texture and is a liquid. It has many lab applications including use as an antifreeze, use as a desiccant, protecting group for carbonyls during chemical synthesis and as a preservative for biological specimens. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.
Specifications
Specifications
| Acidity | 0.005% |
| CAS Max % | 100% |
| Quantity | 6 x 500 mL |
| Packaging | Poly Bottle |
| Boiling Point | 196°C to 199°C |
| Percent Purity | 99% |
| Grade | Reagent |
| Chemical Name or Material | Ethylene Glycol |