missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Glycol, puriss. p.a., Reag. Ph. Eur., ≥99.5%, Honeywell™ Riedel-de Haen™

Catalog No. p-7120431
Change view
Click to view available options
Quantity:
250 mL
1 L
5 L
6 x 1 L
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60044523 250 mL Poly Bottle
60044522 1 L Poly Bottle
60044524 5 L Poly Bottle
60044525 6 x 1 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60044523 Supplier Honeywell Chemicals Supplier No. 03750250ML
Only null left

Puriss. p.a., Reag. Ph. Eur.,≥99.5%

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.

Chemical Identifiers

CAS 107-21-1
Molecular Formula C2H6O2
Molecular Weight (g/mol) 62.068
MDL Number MFCD00002885
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
PubChem CID 174
ChEBI CHEBI:30742
IUPAC Name ethane-1,2-diol
SMILES C(CO)O

Specifications

pH Neutral
Melting Point -12°C
Color Colorless
Density 1.113g/mL at 25°C
Quantity 250 mL
Explosion Limit 0.153
Flash Point 111°C (231.8°F)
Ignition Point 752°F
Packaging Poly Bottle
Physical Form Liquid
Refractive Index n20/D 1.431
Linear Formula HOCH2CH2OH
Vapor Pressure 0.08mmHg (20°C)
UN Number UN3082
Beilstein 505945
Boiling Point 197°C
Formula Weight 62.07g/mol
Percent Purity ≥99.5%
Grade puriss. p.a.
Chemical Name or Material Ethylene Glycol, Reag. Ph. Eur.
Show More Show Less
Hazard Statement P301 + P312 + P330
Precautionary Statement H302-H373
DOTInformation 9/PGIII
EINECSNumber 203-473-3
RTECSNumber KW2975000