missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylene Glycol, Purified, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Boiling Point | 196°C to 199°C |
| Quantity | 1 L |
| Packaging | Poly Bottle |
| Grade | Purified |
| Chemical Name or Material | Ethylene Glycol |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.