missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Glycol, Purified (1,2-Ethanediol; Ethylene Alcohol), Reagents

Catalog No. 01337252
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
15 gal.
200 L
Packaging:
Cubitainer
Glass Bottle
HDPE Winpak
Poly Bottle
Poly Drum
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01337252 500 mL Glass Bottle
01337258 15 gal. Poly Drum
01337257 1 L Glass Bottle
01337256 200 L Poly Drum
01337255 20 L HDPE Winpak
01337254 4 L Poly Bottle
01337253 4 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 01337252 Supplier Labchem Inc Supplier No. C2201500500C
Only null left

Ethylene Glycol, Purified (1,2-Ethanediol; Ethylene Alcohol), Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 107-21-1
Molecular Formula C2H6O2
Molecular Weight (g/mol) 62.07
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym 1,2-Ethanediol, Monoethylene Glycol, Antifreeze
IUPAC Name ethane-1,2-diol
SMILES OCCO

Specifications

Melting Point -13°C
Color Colorless
Quantity 500 mL
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 1.1135
Viscosity Viscous
Boiling Point 198°C
Grade Pure
Chemical Name or Material Ethylene Glycol, Purified (1,2-Ethanediol, Ethylene Alcohol)