missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Glycol Monopropyl Ether 98.0+%, TCI America™

Catalog No. p-7129042
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
E051425ML 25 mL
E0514500ML 500 mL
2 options

Catalog No. E051425ML

Supplier: TCI America E051425ML

Only null left

Chemical Identifiers

CAS 2807-30-9
Molecular Formula C5H12O2
Molecular Weight (g/mol) 104.149
MDL Number MFCD00072645
InChI Key YEYKMVJDLWJFOA-UHFFFAOYSA-N
Synonym ethanol, 2-propoxy, ethylene glycol monopropyl ether, propyl cellosolve, propoxyethanol, propyl glycol, 2-propoxyethan-1-ol, ektasolve ep, 2-propyloxy ethanol, n-propoxyethanol, ethylene glycol mono-n-propyl ether
PubChem CID 17756
IUPAC Name 2-propoxyethanol
SMILES CCCOCCO

Specifications

Color Colorless
Boiling Point 151°C
Quantity 25 mL
UN Number 1992
Formula Weight 104.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethylene Glycol Monopropyl Ether
EINECSNumber (2)-2424&(7)-0097
RTECSNumber KM2800000
TSCA Yes