accessibility menu, dialog, popup

UserName

H3COOH

CAS RN 2807-30-9

IUPAC Name: 2-propoxyethan-1-ol
Synonyms: ethylene glycol monopropyl ether
Molecular Weight (g/mol): 104.15
Molecular Formula: C5H12O2
InChi Key: YEYKMVJDLWJFOA-UHFFFAOYSA-N
SMILES: CCCOCCO
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)