missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylene Glycol Monohexyl Ether 98.0+%, TCI America™
$72.15 - $229.74
Chemical Identifiers
| CAS | 112-25-4 |
|---|---|
| Molecular Formula | C8H18O2 |
| Molecular Weight (g/mol) | 146.23 |
| MDL Number | MFCD00045997 |
| InChI Key | UPGSWASWQBLSKZ-UHFFFAOYSA-N |
| Synonym | Hexyl Cellosolve, 2-(Hexyloxy)ethanol |
| PubChem CID | 8170 |
| IUPAC Name | 2-(hexyloxy)ethan-1-ol |
| SMILES | CCCCCCOCCO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
H034325ML
|
TCI America
H034325ML |
25 mL |
Each for $72.15
|
|
|||||
|
H0343500ML
|
TCI America
H0343500ML |
500 mL |
Each for $229.74
|
|
|||||
Chemical Identifiers
| 112-25-4 | |
| 146.23 | |
| UPGSWASWQBLSKZ-UHFFFAOYSA-N | |
| 8170 | |
| CCCCCCOCCO |
| C8H18O2 | |
| MFCD00045997 | |
| Hexyl Cellosolve, 2-(Hexyloxy)ethanol | |
| 2-(hexyloxy)ethan-1-ol |
Specifications
| 112-25-4 | |
| C8H18O2 | |
| 2922 | |
| -45°C | |
| Hexyl Cellosolve, 2-(Hexyloxy)ethanol | |
| CCCCCCOCCO | |
| 146.23 | |
| 8170 | |
| ≥98.0% (GC) |
| Liquid | |
| MFCD00045997 | |
| Yellow | |
| 205°C | |
| UPGSWASWQBLSKZ-UHFFFAOYSA-N | |
| 2-(hexyloxy)ethan-1-ol | |
| 25 mL | |
| 146.23 | |
| Ethylene Glycol Monohexyl Ether |
Safety and Handling
EINECSNumber : (2)-2424&(7)-0097
RTECSNumber : KL2450000
TSCA : Yes