missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Glycol Monohexyl Ether 98.0+%, TCI America™

Catalog No. H0343500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0343500ML 500 mL
H034325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H0343500ML Supplier TCI America Supplier No. H0343500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 112-25-4
Molecular Formula C8H18O2
Molecular Weight (g/mol) 146.23
MDL Number MFCD00045997
InChI Key UPGSWASWQBLSKZ-UHFFFAOYSA-N
Synonym Hexyl Cellosolve, 2-(Hexyloxy)ethanol
PubChem CID 8170
IUPAC Name 2-(hexyloxy)ethan-1-ol
SMILES CCCCCCOCCO

Specifications

Physical Form Liquid
UN Number 2922
Color Yellow
Melting Point -45°C
Boiling Point 205°C
Quantity 500 mL
Formula Weight 146.23
Percent Purity ≥98.0% (GC)
Chemical Name or Material Ethylene Glycol Monohexyl Ether
EINECSNumber (2)-2424&(7)-0097
RTECSNumber KL2450000
TSCA Yes