missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylene Glycol Mono-m-tolyl Ether 98.0+%, TCI America™
$260.97 - $889.16
Chemical Identifiers
| CAS | 13605-19-1 |
|---|---|
| Molecular Formula | C9H12O2 |
| Molecular Weight (g/mol) | 152.193 |
| MDL Number | MFCD00014035 |
| InChI Key | FDBXUXVQIOQYIX-UHFFFAOYSA-N |
| Synonym | 2-3-methylphenoxy ethanol, 2-m-tolyloxy ethanol, ethanol, 2-3-methylphenoxy, ethylene glycol mono-m-tolyl ether, m-cresoxyethanol, 2-3-tolyloxy ethanol, ethanol, 2-m-tolyloxy, 2-3'-methylphenoxy ethanol, ethylene glycol m-cresyl ether, 2-3-methylphenoxy ethan-1-ol |
| PubChem CID | 83611 |
| IUPAC Name | 2-(3-methylphenoxy)ethanol |
| SMILES | CC1=CC(=CC=C1)OCCO |
Chemical Identifiers
| 13605-19-1 | |
| 152.193 | |
| FDBXUXVQIOQYIX-UHFFFAOYSA-N | |
| 83611 | |
| CC1=CC(=CC=C1)OCCO |
| C9H12O2 | |
| MFCD00014035 | |
| 2-3-methylphenoxy ethanol, 2-m-tolyloxy ethanol, ethanol, 2-3-methylphenoxy, ethylene glycol mono-m-tolyl ether, m-cresoxyethanol, 2-3-tolyloxy ethanol, ethanol, 2-m-tolyloxy, 2-3'-methylphenoxy ethanol, ethylene glycol m-cresyl ether, 2-3-methylphenoxy ethan-1-ol | |
| 2-(3-methylphenoxy)ethanol |
Specifications
| 13605-19-1 | |
| 146°C | |
| MFCD00014035 | |
| 2-3-methylphenoxy ethanol, 2-m-tolyloxy ethanol, ethanol, 2-3-methylphenoxy, ethylene glycol mono-m-tolyl ether, m-cresoxyethanol, 2-3-tolyloxy ethanol, ethanol, 2-m-tolyloxy, 2-3'-methylphenoxy ethanol, ethylene glycol m-cresyl ether, 2-3-methylphenoxy ethan-1-ol | |
| CC1=CC(=CC=C1)OCCO | |
| 152.193 | |
| 152.19 | |
| Liquid |
| Colorless | |
| C9H12O2 | |
| 5 mL | |
| FDBXUXVQIOQYIX-UHFFFAOYSA-N | |
| 2-(3-methylphenoxy)ethanol | |
| 83611 | |
| ≥98.0% (GC) | |
| Ethylene Glycol Mono-m-tolyl Ether |
Safety and Handling
EINECSNumber : (3)-2777
TSCA : Yes