missing translation for 'onlineSavingsMsg'
Learn More

Ethylene glycol ethyl methyl ether, 97%, stab. with 0.01% BHT, Thermo Scientific Chemicals

$93.40 - $347.77

Chemical Identifiers

CAS 5137-45-1
Molecular Formula C5H12O2
MDL Number MFCD00048580
Synonym 2,5-dioxaheptane, ethane, 1-ethoxy-2-methoxy, ethyl 2-methoxyethyl ether
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2081909
View Documents
Thermo Scientific Chemicals
B2081909
10 g
Each for $93.40
Only null left
 
AAB2081918
View Documents
Thermo Scientific Chemicals
B2081918
50 g
Each for $347.77
Only null left
 
Description

Description

It is employed in the reaction of metal salts of thymine and uracil with tetra-acetyl-α-glucopyranosyl bromide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the reaction of metal salts of thymine and uracil with tetra-acetyl-α-glucopyranosyl bromide.

Solubility
Fully miscible in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

5137-45-1
C5H12O2
MFCD00048580
2,5-dioxaheptane, ethane, 1-ethoxy-2-methoxy, ethyl 2-methoxyethyl ether

Specifications

5137-45-1
100°C to 102°C
C5H12O2
1.387
10 g
2,5-dioxaheptane, ethane, 1-ethoxy-2-methoxy, ethyl 2-methoxyethyl ether
104.15
Ethylene glycol ethyl methyl ether, stabilized with 0.01% BHT
0.85
2°C (36°F)
CH3OCH2CH2OCH2CH3
MFCD00048580
UN3271
Fully miscible in water.
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H225-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHERS, N.O.S.

EINECSNumber : 225-893-6

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only