accessibility menu, dialog, popup

UserName

H3COOCH3

CAS RN 5137-45-1

IUPAC Name: 1-ethoxy-2-methoxyethane
Synonyms: 1-ethoxy-2-methoxyethane
Molecular Weight (g/mol): 104.15
Molecular Formula: C5H12O2
InChi Key: CAQYAZNFWDDMIT-UHFFFAOYSA-N
SMILES: CCOCCOC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)