missing translation for 'onlineSavingsMsg'
Learn More

Ethylene glycol ethyl methyl ether, 97%, stab. with 0.01% BHT, Thermo Scientific Chemicals

Catalog No. AAB2081918
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2081918 50 g
AAB2081909 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2081918 Supplier Thermo Scientific Chemicals Supplier No. B2081918
Get Up to $1,800 in Chemicals with Purchase
View all promotions

It is employed in the reaction of metal salts of thymine and uracil with tetra-acetyl-α-glucopyranosyl bromide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the reaction of metal salts of thymine and uracil with tetra-acetyl-α-glucopyranosyl bromide.

Solubility
Fully miscible in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 5137-45-1
Molecular Formula C5H12O2
MDL Number MFCD00048580
Synonym 2,5-dioxaheptane, ethane, 1-ethoxy-2-methoxy, ethyl 2-methoxyethyl ether

Specifications

Density 0.85
Boiling Point 100°C to 102°C
Flash Point 2°C (36°F)
Refractive Index 1.387
Linear Formula CH3OCH2CH2OCH2CH3
Quantity 50 g
UN Number UN3271
Solubility Information Fully miscible in water.
Formula Weight 104.15
Percent Purity 97%
Chemical Name or Material Ethylene glycol ethyl methyl ether, stabilized with 0.01% BHT
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHERS, N.O.S.
EINECSNumber 225-893-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only