missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Glycol Dimethacrylate (stabilized with HQ) 97.0+%, TCI America™

Catalog No. E0102500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0102500ML 500 mL
E010225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E0102500ML Supplier TCI America Supplier No. E0102500ML

Chemical Identifiers

CAS 97-90-5
Molecular Formula C10H14O4
Molecular Weight (g/mol) 198.22
MDL Number MFCD00008590
InChI Key STVZJERGLQHEKB-UHFFFAOYSA-N
Synonym ethylene glycol dimethacrylate, ethylene dimethacrylate, glycol dimethacrylate, diglycol dimethacrylate, ethanediol dimethacrylate, ethylenedimethyacrylate, ethylene methacrylate, ethyldiol metacrylate, 1,2-bis methacryloyloxy ethane, ethylene glycol bis methacrylate
PubChem CID 7355
ChEBI CHEBI:53436
IUPAC Name 2-[(2-methylprop-2-enoyl)oxy]ethyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCCOC(=O)C(C)=C

Specifications

Melting Point -40°C
Color Colorless
Boiling Point 235°C
Quantity 500 mL
Formula Weight 198.22
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethylene Glycol Dimethacrylate (stabilized with HQ)
EINECSNumber (2)-1056&(2)-1059
RTECSNumber OZ4400000
TSCA Yes