missing translation for 'onlineSavingsMsg'
Learn More

Ethylene glycol dimethacrylate, 98%, stab. with 100ppm 4-methoxyphenol

Catalog No. p-7059189
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
4 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4415118 50 g
AA4415130 250 g
AA44151A1 1 kg
AA44151A6 4 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA4415118 Supplier Thermo Scientific Chemicals Supplier No. 04415118

Crosslinking monomer | CAS: 97-90-5 | C10H14O4 | 198.22 g/mol

Ethylene glycol dimethacrylate is used as a functional monomer for polymers and as a cross linking agent between the molecular chains of polymers and elastomers. It is also used in free radical copolymer cross linking reactions. It acts as an intermediate in the production of hydroxyapatite and poly methyl methacrylate composites.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 97-90-5
Molecular Formula C10H14O4
Molecular Weight (g/mol) 198.22
MDL Number MFCD00008590
InChI Key STVZJERGLQHEKB-UHFFFAOYSA-N
Synonym ethylene glycol dimethacrylate, ethylene dimethacrylate, glycol dimethacrylate, diglycol dimethacrylate, ethanediol dimethacrylate, ethylenedimethyacrylate, ethylene methacrylate, ethyldiol metacrylate, 1,2-bis methacryloyloxy ethane, ethylene glycol bis methacrylate
PubChem CID 7355
ChEBI CHEBI:53436
IUPAC Name 2-(2-methylprop-2-enoyloxy)ethyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCCOC(=O)C(C)=C

Specifications

Color Colorless to Yellow
Density 1.051 g/mL
Boiling Point 83°C to 85°C (1 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.454
Linear Formula H2C=C(CH3)CO2CH2CH2OCOC(CH3)=CH2
Quantity 50 g
Beilstein 1776663
Sensitivity Light sensitive
Solubility Information Soluble in water.
Formula Weight 198.21
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethylene glycol dimethacrylate, Stabilized with 100ppm 4-methoxyphenol
Show More Show Less
Hazard Category H317-H335
Hazard Statement GHS H Statement
H317-H335
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P261-P271-P272-P280g-P302+P352-P304+P340-P312-P333+P313-P363-P501c
EINECSNumber 202-617-2
RTECSNumber OZ4400000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only