missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Glycol Dibenzyl Ether 95.0+%, TCI America™

Catalog No. p-7126066
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D01605ML 5 mL
D016025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D01605ML Supplier TCI America Supplier No. D01605ML

Chemical Identifiers

CAS 622-22-0
Molecular Formula C16H18O2
Molecular Weight (g/mol) 242.32
MDL Number MFCD00030016
InChI Key FPFHYKOBYMYVAN-UHFFFAOYSA-N
Synonym 1,2-Bis(benzyloxy)ethane, Dibenzyl Cellosolve, Dibenzyl Glycol, 1,2-Dibenzyloxyethane
PubChem CID 222542
IUPAC Name {[2-(benzyloxy)ethoxy]methyl}benzene
SMILES C(COCC1=CC=CC=C1)OCC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 210°C
Quantity 5 mL
Formula Weight 242.32
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethylene Glycol Dibenzyl Ether