missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Glycol (Certified), Fisher Chemical™

Purity is guaranteed to meet published maximum limits of impurities

$576.08 - $576.08

Chemical Identifiers

CAS 107-21-1
Molecular Formula C2H6O2
Molecular Weight (g/mol) 62.068
MDL Number MFCD00002885
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
PubChem CID 174
ChEBI CHEBI:30742
IUPAC Name ethane-1,2-diol
SMILES C(CO)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
E178-4
SDS Accessories View Documents
Fisher Chemical
E1784
4 L Glass Bottle
Each for $576.08
Case of 4 Each for $2,304.32
Only null left
 
Description

Description

Specifications

Chemical Identifiers

107-21-1
62.068
LYCAIKOWRPUZTN-UHFFFAOYSA-N
174
ethane-1,2-diol
C2H6O2
MFCD00002885
ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
CHEBI:30742
C(CO)O

Specifications

5.5 to 7.5
107-21-1
0.01% max.
1.113g/cm³
4 L
Pass Test
Viscous Liquid
C2H6O2
MFCD00002885
21 cP at 20°C
ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
C(CO)O
62.068
CHEBI:30742
Certified
-13°C
Colorless
>500°C
196°C
116°C (240.8°F)
Glass Bottle
1.112 to 1.115 (at 25°C)
HOCH2CH2OH
0.12mmHg at 20°C
0.2% max.
LYCAIKOWRPUZTN-UHFFFAOYSA-N
ethane-1,2-diol
174
62.07
Ethylene Glycol
Videos
Safety and Handling

Safety and Handling

WARNING!

Emergency Overview
Harmful if swallowed. May cause skin, eye, and respiratory tract irritation. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur.

NFPA

Health:2
Flammability:1
Instability:0

SDS
missing translation for 'documents'

missing translation for 'documents'