missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylene Glycol Bis(propionitrile) Ether 97.0+%, TCI America™
$255.38 - $736.30
Chemical Identifiers
| CAS | 3386-87-6 |
|---|---|
| Molecular Formula | C8H12N2O2 |
| Molecular Weight (g/mol) | 168.196 |
| MDL Number | MFCD00019871 |
| InChI Key | VTHRQKSLPFJQHN-UHFFFAOYSA-N |
| PubChem CID | 76921 |
| IUPAC Name | 3-[2-(2-cyanoethoxy)ethoxy]propanenitrile |
| SMILES | C(COCCOCCC#N)C#N |
Chemical Identifiers
| 3386-87-6 | |
| 168.196 | |
| VTHRQKSLPFJQHN-UHFFFAOYSA-N | |
| 3-[2-(2-cyanoethoxy)ethoxy]propanenitrile |
| C8H12N2O2 | |
| MFCD00019871 | |
| 76921 | |
| C(COCCOCCC#N)C#N |
Specifications
| 3386-87-6 | |
| 165°C | |
| MFCD00019871 | |
| 3276 | |
| VTHRQKSLPFJQHN-UHFFFAOYSA-N | |
| 3-[2-(2-cyanoethoxy)ethoxy]propanenitrile | |
| 76921 | |
| ≥97.0% (GC) | |
| Ethylene Glycol Bis(propionitrile) Ether |
| Yellow | |
| C8H12N2O2 | |
| 25 g | |
| 1,2-Bis(2-cyanoethoxy)ethane, 3,3′C-(Ethylenedioxy)dipropionitrile, Ethylene Glycol Bis(2-cyanoethyl) Ether | |
| C(COCCOCCC#N)C#N | |
| 168.196 | |
| 168.20 | |
| Liquid |
Safety and Handling
TSCA : Yes