accessibility menu, dialog, popup

UserName

NOON

CAS RN 3386-87-6

IUPAC Name: 3-[2-(2-cyanoethoxy)ethoxy]propanenitrile
Synonyms: 3-[2-(2-cyanoethoxy)ethoxy]propanenitrile
Molecular Weight (g/mol): 168.2
Molecular Formula: C8H12N2O2
InChi Key: VTHRQKSLPFJQHN-UHFFFAOYSA-N
SMILES: N#CCCOCCOCCC#N
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results