missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Glycol Bis(propionitrile) Ether 97.0+%, TCI America™

Catalog No. E010825G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E010825G 25 g
E0108100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E010825G Supplier TCI America Supplier No. E010825G
Only null left

Chemical Identifiers

CAS 3386-87-6
Molecular Formula C8H12N2O2
Molecular Weight (g/mol) 168.196
MDL Number MFCD00019871
InChI Key VTHRQKSLPFJQHN-UHFFFAOYSA-N
PubChem CID 76921
IUPAC Name 3-[2-(2-cyanoethoxy)ethoxy]propanenitrile
SMILES C(COCCOCCC#N)C#N

Specifications

Color Yellow
Boiling Point 165°C
Quantity 25 g
UN Number 3276
Synonym 1,2-Bis(2-cyanoethoxy)ethane, 3,3′C-(Ethylenedioxy)dipropionitrile, Ethylene Glycol Bis(2-cyanoethyl) Ether
Formula Weight 168.20
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethylene Glycol Bis(propionitrile) Ether