missing translation for 'onlineSavingsMsg'
Learn More

Ethylene glycol-bis(beta-aminoethyl ether)-N,N,N',N'-tetraacetic acid tetrasodium salt, 95%

Catalog No. p-4112115
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC428570100 10 g Glass bottle
AC428570500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC428570100 Supplier Thermo Scientific Chemicals Supplier No. 428570100
Only null left

CAS: 13368-13-3 | C14H20N2Na4O10 | 468.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13368-13-3
Molecular Formula C14H20N2Na4O10
Molecular Weight (g/mol) 468.28
MDL Number MFCD00274420
InChI Key BAOGCDPNNOBWQZ-UHFFFAOYSA-J
Synonym egta tetrasodium, 3,12-bis carboxymethyl-6,9-dioxa-3,12-diazatetradecane-1,14-dioic acid, tetrasodium salt, ethylene glycol-bis beta-aminoethyl ether-n,n,n',n'-tetraacetic acid tetrasodium salt, egta tetrasodium salt, egtazic acid tetrasodium, ethylene-bis oxyethylenenitrilo tetraacetic acid tetrasodium, 2,2',2,2'-ethylenebis oxyethylenenitrilo tetraacetic acid tetrasodium salt, 6,9-dioxa-3,12-diazatetradecanedioicacid,3,12-bis carboxymethyl-,sodium salt 1:4, tetrasodium 3,12-bis carboxylatomethyl-6,9-dioxa-3,12-diazatetradecane-1,14-dioate, ethylene glycol-bis, a-aminoethyl ether-n,n,n inverted exclamation marka,n inverted exclamation marka-tetraacetic acid tetrasodium salt
PubChem CID 16219327
IUPAC Name tetrasodium;2-[2-[2-[2-[bis(carboxylatomethyl)amino]ethoxy]ethoxy]ethyl-(carboxylatomethyl)amino]acetate
SMILES C(COCCOCCN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)[O-])CC(=O)[O-].[Na+].[Na+].[Na+].[Na+]

Specifications

Melting Point 54.0°C to 56.0°C
Assay Percent Range 18.46 to 20.82% (ICP on dried sample)
Packaging Glass bottle
Quantity 10 g
Solubility Information Solubility in water: soluble.
Formula Weight 468.28
Percent Purity 95%
Chemical Name or Material Ethylene glycol-bis(β-aminoethyl ether)-N,N,N',N'-tetraacetic acid tetrasodium salt
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only