accessibility menu, dialog, popup

UserName

OONOOOONOOOONaNaNaNa

CAS RN 13368-13-3

IUPAC Name: tetrasodium 3,12-bis(carboxylatomethyl)-6,9-dioxa-3,12-diazatetradecanedioate
Synonyms: tetrasodium egta
Molecular Weight (g/mol): 468.28
Molecular Formula: C14H20N2Na4O10
InChi Key: BAOGCDPNNOBWQZ-UHFFFAOYSA-J
SMILES: [Na+].[Na+].[Na+].[Na+].[O-]C(=O)CN(CCOCCOCCN(CC([O-])=O)CC([O-])=O)CC([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results