missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Glycol Bis(4-carboxyphenyl) Ether 95.0+%, TCI America™

Catalog No. E04155G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E04155G 5 g
E04151G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E04155G Supplier TCI America Supplier No. E04155G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3753-05-7
Molecular Formula C16H14O6
Molecular Weight (g/mol) 302.28
MDL Number MFCD00020365
InChI Key VAXBLYWAVAIJJJ-UHFFFAOYSA-N
Synonym 1,2-Bis(4-carboxyphenoxy)ethane
PubChem CID 77358
IUPAC Name 4-[2-(4-carboxyphenoxy)ethoxy]benzoic acid
SMILES C1=CC(=CC=C1C(=O)O)OCCOC2=CC=C(C=C2)C(=O)O

Specifications

Color White
Quantity 5 g
Formula Weight 302.28
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Ethylene Glycol Bis(4-carboxyphenyl) Ether