missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylene Glycol Bis(3-mercaptopropionate) (Purified) 98.0+%, TCI America™
$292.65 - $874.24
Chemical Identifiers
| CAS | 22504-50-3 |
|---|---|
| Molecular Formula | C8H14O4S2 |
| Molecular Weight (g/mol) | 238.316 |
| MDL Number | MFCD00053520 |
| InChI Key | HAQZWTGSNCDKTK-UHFFFAOYSA-N |
| Synonym | Ethylene Bis(3-mercaptopropionate), Glycol Bis(3-mercaptopropionate) |
| PubChem CID | 89735 |
| IUPAC Name | 2-(3-sulfanylpropanoyloxy)ethyl 3-sulfanylpropanoate |
| SMILES | C(CS)C(=O)OCCOC(=O)CCS |
Chemical Identifiers
| 22504-50-3 | |
| 238.316 | |
| HAQZWTGSNCDKTK-UHFFFAOYSA-N | |
| 89735 | |
| C(CS)C(=O)OCCOC(=O)CCS |
| C8H14O4S2 | |
| MFCD00053520 | |
| Ethylene Bis(3-mercaptopropionate), Glycol Bis(3-mercaptopropionate) | |
| 2-(3-sulfanylpropanoyloxy)ethyl 3-sulfanylpropanoate |
Specifications
| 22504-50-3 | |
| C8H14O4S2 | |
| 25 g | |
| HAQZWTGSNCDKTK-UHFFFAOYSA-N | |
| 2-(3-sulfanylpropanoyloxy)ethyl 3-sulfanylpropanoate | |
| 89735 | |
| ≥98.0% (T) | |
| Ethylene Glycol Bis(3-mercaptopropionate) (Purified) |
| Yellow | |
| MFCD00053520 | |
| Ethylene Bis(3-mercaptopropionate), Glycol Bis(3-mercaptopropionate) | |
| C(CS)C(=O)OCCOC(=O)CCS | |
| 238.316 | |
| 238.32 | |
| Liquid |
Safety and Handling
TSCA : Yes