missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Glycol Bis(3-aminopropyl) Ether 98.0+%, TCI America™

Catalog No. p-7126962
Change view
Click to view available options
Quantity:
5 mL
25 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D18405ML 5 mL
D184025ML 25 mL
D1840500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D18405ML Supplier TCI America Supplier No. D18405ML
Only null left

Chemical Identifiers

CAS 2997-01-5
Molecular Formula C8H22N2O2
Molecular Weight (g/mol) 178.28
MDL Number MFCD00059938
InChI Key POTQBGGWSWSMCX-UHFFFAOYSA-P
Synonym di 3-aminopropoxy ethane, 1-propanamine, 3,3'-1,2-ethanediylbis oxy bis, 1,2-bis 3-aminopropoxy ethane, 3,3'-ethane-1,2-diylbis oxy bis propan-1-amine, 3,3'-ethylenedioxy bispropylamine, 3,3'-ethylenedioxybis propylamine, propylamine, 3,3'-ethylenedioxy bis, ethylene glycol bis 3-aminopropyl ether, 1,10-diamino-4,7-dioxadecane, 3-2-3-aminopropoxy ethoxy propan-1-amine
PubChem CID 76342
IUPAC Name 3-[2-(3-aminopropoxy)ethoxy]propan-1-amine
SMILES C(CN)COCCOCCCN

Specifications

Color Yellow
Boiling Point 96°C
Quantity 5 mL
UN Number 3267
Formula Weight 176.26
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Ethylene Glycol Bis(3-aminopropyl) Ether
RTECSNumber UI2800000
TSCA Yes