accessibility menu, dialog, popup

UserName

H3NOONH3

CAS RN 2997-01-5

IUPAC Name: 3-[2-(3-azaniumylpropoxy)ethoxy]propan-1-aminium
Synonyms: 3-[2-(3-ammoniopropoxy)ethoxy]propan-1-aminium
Molecular Weight (g/mol): 178.28
Molecular Formula: C8H22N2O2
InChi Key: POTQBGGWSWSMCX-UHFFFAOYSA-P
SMILES: [NH3+]CCCOCCOCCC[NH3+]
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results