missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylene Glycol Bis(3-aminopropyl) Ether 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 96°C |
| Quantity | 5 mL |
| UN Number | 3267 |
| Formula Weight | 176.26 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Liquid |
| Chemical Name or Material | Ethylene Glycol Bis(3-aminopropyl) Ether |
Safety and Handling
| RTECSNumber | UI2800000 |
| TSCA | Yes |