missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Glycol Bis(2-nitrophenyl) Ether 98.0+%, TCI America™

Catalog No. p-7129275
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E10061G 1 g
E10065G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E10061G Supplier TCI America Supplier No. E10061G
Only null left

Chemical Identifiers

CAS 51661-19-9
Molecular Formula C14H12N2O6
Molecular Weight (g/mol) 304.26
MDL Number MFCD00024226
InChI Key DEHTVRKGDUCXRF-UHFFFAOYSA-N
Synonym 1,2-bis o-nitrophenoxy ethane, 1,2-bis 2-nitrophenoxy ethane, 1-nitro-2-2-2-nitrophenoxy ethoxy benzene, ethylene glycol bis 2-nitrophenyl ether, 1,1'-ethane-1,2-diylbis oxy bis 2-nitrobenzene, bis 2-nitrophenoxy ethane, acmc-20a59z, 1,2-di o-nitrophenoxy ethane, 2,2'-ethylenedioxy bis 1-nitrobenzene, 2-nitro-1-2-2-nitrophenoxy ethoxy benzene
PubChem CID 394580
IUPAC Name 1-nitro-2-[2-(2-nitrophenoxy)ethoxy]benzene
SMILES [O-][N+](=O)C1=CC=CC=C1OCCOC1=CC=CC=C1[N+]([O-])=O

Specifications

Melting Point 170°C
Color Yellow
Quantity 1 g
Formula Weight 304.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethylene Glycol Bis(2-nitrophenyl) Ether